Gene Transcripts

Gene Symbol
ELMO2
Entrez Gene ID
63916
Gene Name
engulfment and cell motility 2
Gene Aliases
CED-12, CED12, Ced-12A, ELMO-2
Chromosome Location
Chr. 20: 46366045 - 46407051 on Build GRCh38
UniGene ID
Hs.210469
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318253.1 NP_001305182.1 7, 8 1129
NM_133171.4 NP_573403.1 7, 8 1061
NM_182764.2 NP_877496.1 6, 7 957
XM_005260496.3 XP_005260553.1 7, 8 626
XM_005260498.3 XP_005260555.1 6, 7 988
XM_005260499.3 XP_005260556.1 6, 7 959
XM_005260500.3 XP_005260557.1 5, 6 487
XM_006723854.3 XP_006723917.1 6, 7 560
GenBank mRNA AB058737.1 - 6, 7 572
AF398886.1 - 5, 6 500
AF417861.1 - 5, 6 432
AK057032.1 - 7, 8 585
AK299318.1 - 8, 9 636
AY947527.1 - 1, 2 67
BC000143.4 - 6, 7 637 3047992
BX648819.1 - 6, 7 538

View Full Product Details