Gene Transcripts
- Gene Symbol
- CREB3L2
- Entrez Gene ID
- 64764
- Gene Name
- cAMP responsive element binding protein 3 like 2
- Gene Aliases
- BBF2H7
- Chromosome Location
- Chr. 7: 137874979 - 138002101 on Build GRCh38
- UniGene ID
- Hs.490273
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001318246.1 |
NP_001305175.1
|
9, 10 |
1130 |
|
NM_194071.3 |
NP_919047.2
|
9, 10 |
1543 |
|
XM_005250541.4 |
XP_005250598.1
|
9, 10 |
1922 |
|
XM_017012540.1 |
XP_016868029.1
|
10, 11 |
2945 |
|
XM_017012541.1 |
XP_016868030.1
|
10, 11 |
1508 |
|
GenBank mRNA |
AJ549092.1 |
- |
9, 10 |
1496 |
|
AJ549387.1 |
- |
9, 10 |
1496 |
|
AK131517.1 |
- |
9, 10 |
1509 |
|
AK293048.1 |
- |
9, 10 |
1475 |
|
BC110813.1 |
- |
9, 10 |
1479 |
3923538 |
BX649143.1 |
- |
9, 10 |
1135 |
|
CR749563.1 |
- |
8, 9 |
1223 |
|
View Full Product Details