Gene Transcripts
- Gene Symbol
- MRPS9
- Entrez Gene ID
- 64965
- Gene Name
- mitochondrial ribosomal protein S9
- Gene Aliases
- MRP-S9, RPMS9, S9mt
- Chromosome Location
- Chr. 2: 105038025 - 105099960 on Build GRCh38
- UniGene ID
- Hs.590900
- Species
- Homo sapiens
- Gene Symbol
- UTAT33
- Entrez Gene ID
- 101927492
- Gene Name
- uncharacterized LOC101927492
- Gene Aliases
- Chromosome Location
- Chr. 2: 105097052 - 105102944 on Build GRCh38
- UniGene ID
- Hs.469537
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_110227.1 |
|
4 |
436 |
|
GenBank mRNA |
AI381531.1 |
- |
1 |
137 |
|
CR736606.1 |
- |
4 |
437 |
|
View Full Product Details