Gene Transcripts

Gene Symbol
FASTKD3
Entrez Gene ID
79072
Gene Name
FAST kinase domains 3
Gene Aliases
Chromosome Location
Chr. 5: 7859159 - 7869037 on Build GRCh38
UniGene ID
Hs.653162
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024091.3 NP_076996.2 2 813
XM_006714498.1 XP_006714561.1 2 724
GenBank mRNA AK026927.1 - 2 778
BC001295.1 - 1 305
BC113563.1 - 1 728 8322615
BC143517.1 - 1 728

View Full Product Details