Gene Transcripts

Gene Symbol
WDR77
Entrez Gene ID
79084
Gene Name
WD repeat domain 77
Gene Aliases
HKMT1069, MEP-50, MEP50, Nbla10071, p44, p44/Mep50
Chromosome Location
Chr. 1: 111439890 - 111449308 on Build GRCh38
UniGene ID
Hs.204773
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317062.1 NP_001303991.1 3, 4 580
NM_001317063.1 NP_001303992.1 3, 4 580
NM_024102.3 NP_077007.1 3, 4 580
GenBank mRNA AB073603.1 - 3, 4 528
AF052183.1 - 3, 4 466
AF478464.1 - 3, 4 441
AK022860.1 - 3, 4 510
AK223189.1 - 3, 4 510
AL557974.3 - 3, 4 496
AY225316.1 - 3, 4 466
BC001679.2 - 3, 4 501 2822173
BC006477.1 - 3, 4 501 2822173
BC009411.1 - 3, 4 513 3614223
BC011778.2 - 3, 4 520 3941878
BC016946.1 - 3, 4 480 3845908
BI195029.1 - 2, 3 252

View Full Product Details