Gene Transcripts

Gene Symbol
WDR77
Entrez Gene ID
79084
Gene Name
WD repeat domain 77
Gene Aliases
HKMT1069, MEP-50, MEP50, Nbla10071, p44, p44/Mep50
Chromosome Location
Chr. 1: 111439890 - 111449308 on Build GRCh38
UniGene ID
Hs.204773
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317062.1 NP_001303991.1 9, 10 1034
NM_001317064.1 NP_001303993.1 7, 8 806
NM_024102.3 NP_077007.1 9, 10 998
NR_133654.1 8, 9 856
GenBank mRNA AB073603.1 - 9, 10 946
AB074171.1 - 4, 5 310
AF052183.1 - 9, 10 884
AF478464.1 - 9, 10 859
AK022860.1 - 9, 10 928
AK223189.1 - 9, 10 928
AK296163.1 - 8, 9 800
AK298179.1 - 7, 8 750
AY225316.1 - 9, 10 884
BC001679.2 - 9, 10 919 2822173
BC006477.1 - 9, 10 919 2822173
BC009411.1 - 9, 10 931 3614223
BC011778.2 - 9, 10 938 3941878
BC016946.1 - 9, 10 898 3845908

View Full Product Details