Gene Transcripts
- Gene Symbol
- ZSCAN5A
- Entrez Gene ID
- 79149
- Gene Name
- zinc finger and SCAN domain containing 5A
- Gene Aliases
- ZNF495, ZSCAN5
- Chromosome Location
- Chr. 19: 56221303 - 56368383 on Build GRCh38
- UniGene ID
- Hs.177688
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001322061.1 |
NP_001308990.1
|
4, 5 |
921 |
|
NM_001322062.1 |
NP_001308991.1
|
3, 4 |
689 |
|
NM_001322064.1 |
NP_001308993.1
|
3, 4 |
858 |
|
NM_001322065.1 |
NP_001308994.1
|
3, 4 |
733 |
|
NM_001322066.1 |
NP_001308995.1
|
3, 4 |
928 |
|
NM_001322067.1 |
NP_001308996.1
|
3, 4 |
632 |
|
NM_001322068.1 |
NP_001308997.1
|
3, 4 |
757 |
|
NM_001322069.1 |
NP_001308998.1
|
3, 4 |
928 |
|
NM_001322070.1 |
NP_001308999.1
|
3, 4 |
803 |
|
NM_001322072.1 |
NP_001309001.1
|
4, 5 |
1080 |
|
NM_001322073.1 |
NP_001309002.1
|
3, 4 |
733 |
|
NM_001322074.1 |
NP_001309003.1
|
3, 4 |
860 |
|
NM_001322075.1 |
NP_001309004.1
|
3, 4 |
928 |
|
NM_001322076.1 |
NP_001309005.1
|
3, 4 |
803 |
|
NM_024303.2 |
NP_077279.1
|
3, 4 |
814 |
|
XM_005259254.1 |
XP_005259311.1
|
3, 4 |
689 |
|
XM_006723377.3 |
XP_006723440.1
|
2, 3 |
973 |
|
XM_011527308.1 |
XP_011525610.1
|
2, 3 |
498 |
|
XM_017027298.1 |
XP_016882787.1
|
2, 3 |
958 |
|
XM_017027299.1 |
XP_016882788.1
|
2, 3 |
1055 |
|
GenBank mRNA |
AK098700.1 |
- |
2, 3 |
446 |
|
AK223485.1 |
- |
1, 2 |
511 |
|
BC002636.1 |
- |
2, 3 |
550 |
3606994 |
BC043232.1 |
- |
4, 5 |
1085 |
5295665 |
View Full Product Details