Gene Transcripts

Gene Symbol
OR13A1
Entrez Gene ID
79290
Gene Name
olfactory receptor family 13 subfamily A member 1
Gene Aliases
Chromosome Location
Chr. 10: 45302294 - 45315608 on Build GRCh38
UniGene ID
Hs.532435
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004297.2 NP_001004297.2 4 884
XM_011540147.2 XP_011538449.1 3 1351
XM_017016632.1 XP_016872121.1 4 4270
XM_017016633.1 XP_016872122.1 3 4412
XM_017016634.1 XP_016872123.1 3 4657
GenBank mRNA AK131555.1 - 4 884
BC104853.1 - 2 687 8143856
BC104855.1 - 2 687 8143858
BC143622.1 - 2 687 9052135

View Full Product Details