Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_024552.2 | NP_078828.2 | 10 | 1145 | |
XM_011528290.2 | XP_011526592.1 | 10 | 1181 | ||
XM_011528291.2 | XP_011526593.1 | 10 | 1298 | ||
XM_011528292.2 | XP_011526594.1 | 11 | 2121 | ||
XM_011528293.2 | XP_011526595.1 | 11 | 1297 | ||
XM_011528294.2 | XP_011526596.1 | 11 | 1265 | ||
XM_011528295.2 | XP_011526597.1 | 9 | 951 | ||
XM_017027303.1 | XP_016882792.1 | 11 | 2153 | ||
XM_017027304.1 | XP_016882793.1 | 12 | 1376 | ||
XM_017027305.1 | XP_016882794.1 | 9 | 1012 | ||
GenBank mRNA | AK022151.1 | - | 10 | 1105 | |
AK055414.1 | - | 7 | 1682 | ||
AK222621.1 | - | 10 | 1112 | ||
BC009828.2 | - | 9 | 948 | 3936998 |