Gene Transcripts

Gene Symbol
TMEM53
Entrez Gene ID
79639
Gene Name
transmembrane protein 53
Gene Aliases
NET4
Chromosome Location
Chr. 1: 44653247 - 44674608 on Build GRCh38
UniGene ID
Hs.22157
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001300746.1 NP_001287675.1 2 381
NM_001300747.1 NP_001287676.1 3 413
NM_001300748.1 NP_001287677.1 2, 3 410
NM_024587.3 NP_078863.2 3 503
XM_011542138.2 XP_011540440.1 3 632
GenBank mRNA AA490195.1 - 2, 3 205
AK026006.1 - 3 310
AK225405.1 - 3 310
AK295356.1 - 1, 2 103
AK296548.1 - 2 219
AK310016.1 - 3 447
AL832539.1 - 2 215
BC064520.1 - 3 322 5582377
CR457348.1 - 3 286
HY029536.1 - 3 247

View Full Product Details