Gene Transcripts

Gene Symbol
AAGAB
Entrez Gene ID
79719
Gene Name
alpha- and gamma-adaptin binding protein
Gene Aliases
KPPP1, PPKP1, PPKP1A, p34
Chromosome Location
Chr. 15: 67200675 - 67255198 on Build GRCh38
UniGene ID
Hs.254642
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271885.1 NP_001258814.1 2 135
NM_001271886.1 NP_001258815.1 2 396
NM_024666.4 NP_078942.3 2 251
XM_011522020.1 XP_011520322.1 2 501
GenBank mRNA AK021568.1 - 2 155
AK294402.1 - 2 132
AL136715.1 - 2 161
BC047026.1 - 2 174 5534878
BC058886.1 - 2 173 6503238
CR533540.1 - 2 146
DA160293.1 - 2 311
DA688370.1 - 2 251
DC315715.1 - 2 135

View Full Product Details