Gene Transcripts
- Gene Symbol
- DHRS12
- Entrez Gene ID
- 79758
- Gene Name
- dehydrogenase/reductase 12
- Gene Aliases
- SDR40C1
- Chromosome Location
- Chr. 13: 51754856 - 51804206 on Build GRCh38
- UniGene ID
- Hs.266728
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001031719.2 |
NP_001026889.1
|
5, 6 |
488 |
|
NM_001270424.1 |
NP_001257353.1
|
6, 7 |
571 |
|
NM_024705.2 |
NP_078981.1
|
5, 6 |
488 |
|
XM_005266527.3 |
XP_005266584.1
|
5, 6 |
494 |
|
XM_005266528.4 |
XP_005266585.1
|
6, 7 |
615 |
|
XM_011535236.2 |
XP_011533538.1
|
6, 7 |
615 |
|
XM_011535238.1 |
XP_011533540.1
|
5, 6 |
494 |
|
XM_017020750.1 |
XP_016876239.1
|
6, 7 |
615 |
|
XM_017020751.1 |
XP_016876240.1
|
5, 6 |
494 |
|
XM_017020752.1 |
XP_016876241.1
|
5, 6 |
494 |
|
GenBank mRNA |
AK023701.1 |
- |
5, 6 |
483 |
|
BC009825.2 |
- |
5, 6 |
424 |
3942115 |
BC026024.1 |
- |
6, 7 |
504 |
|
View Full Product Details