Gene Transcripts

Gene Symbol
MYH14
Entrez Gene ID
79784
Gene Name
myosin, heavy chain 14, non-muscle
Gene Aliases
DFNA4, DFNA4A, FP17425, MHC16, MYH17, NMHC II-C, NMHC-II-C, PNMHH, myosin
Chromosome Location
Chr. 19: 50188186 - 50310544 on Build GRCh38
UniGene ID
Hs.467142
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077186.1 NP_001070654.1 21, 22 2639
NM_001145809.1 NP_001139281.1 22, 23 2738
NM_024729.3 NP_079005.3 20, 21 2615
XM_006723386.3 XP_006723449.1 21, 22 2700
XM_011527320.2 XP_011525622.1 21, 22 2717
XM_011527321.2 XP_011525623.1 20, 21 2693
XM_011527323.2 XP_011525625.1 21, 22 2707
GenBank mRNA AB111886.1 - 10, 11 1297
AB290169.1 - 21, 22 2708
AK125082.1 - 21, 22 2718
AK307783.1 - 21, 22 2621
AY165122.1 - 20, 21 2615
BC018933.2 - 19, 20 2367

View Full Product Details