Gene Transcripts
- Gene Symbol
- DCAKD
- Entrez Gene ID
- 79877
- Gene Name
- dephospho-CoA kinase domain containing
- Gene Aliases
- Chromosome Location
- Chr. 17: 45023338 - 45061129 on Build GRCh38
- UniGene ID
- Hs.463148
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001128631.2 |
NP_001122103.1
|
3, 4 |
549 |
|
NM_001288654.2 |
NP_001275583.1
|
3, 4 |
685 |
|
NM_001288655.1 |
NP_001275584.1
|
3, 4 |
597 |
|
NM_001321326.1 |
NP_001308255.1
|
3, 4 |
495 |
|
NM_024819.6 |
NP_079095.3
|
3, 4 |
682 |
|
XM_011525262.2 |
XP_011523564.1
|
3, 4 |
714 |
|
GenBank mRNA |
AK290755.1 |
- |
3, 4 |
498 |
|
BC006472.1 |
- |
3, 4 |
571 |
2822306 |
BC006546.2 |
- |
3, 4 |
571 |
2822306 |
BC018132.2 |
- |
2, 3 |
443 |
3903964 |
BC033299.1 |
- |
2, 3 |
582 |
4153431 |
BM910634.1 |
- |
3, 4 |
596 |
|
DR005747.1 |
- |
3, 4 |
597 |
|
View Full Product Details