Gene Transcripts

Gene Symbol
FUZ
Entrez Gene ID
80199
Gene Name
fuzzy planar cell polarity protein
Gene Aliases
FY, NTD
Chromosome Location
Chr. 19: 49806866 - 49817376 on Build GRCh38
UniGene ID
Hs.288800
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001171937.1 NP_001165408.1 9 1089
NM_025129.4 NP_079405.2 10 1197
NR_033269.1 9 1112
XM_006723399.3 XP_006723462.1 10 1175
XM_011527339.1 XP_011525641.1 10 1178
XM_011527340.1 XP_011525642.1 10 1040
XM_011527341.2 XP_011525643.1 10 1104
XM_011527342.1 XP_011525644.1 10 1327
XM_011527343.1 XP_011525645.1 10 1178
XM_011527345.1 XP_011525647.1 10 1136
XM_011527346.1 XP_011525648.1 9 1054
XM_011527347.1 XP_011525649.1 11 1094
XM_017027319.1 XP_016882808.1 10 1037
XM_017027320.1 XP_016882809.1 10 1178
XM_017027321.1 XP_016882810.1 9 1051
XM_017027322.1 XP_016882811.1 10 1037
XM_017027323.1 XP_016882812.1 9 1071
GenBank mRNA AK026341.1 - 10 1171
AK057964.1 - 9 992
AK315445.1 - 10 1175
BC004445.1 - 10 1098 2819805
BC010092.2 - 9 953 3354417
BC016793.2 - 10 1098 2819805

View Full Product Details