Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001126340.2 | NP_001119812.1 | 4 | 896 | |
NM_001271818.1 | NP_001258747.1 | 4 | 919 | ||
NM_001271819.1 | NP_001258748.1 | 3 | 658 | ||
NM_032831.3 | NP_116220.1 | 3 | 787 | ||
GenBank mRNA | AF258552.1 | - | 1 | 74 | |
AK027639.1 | - | 3 | 741 | ||
AK056497.1 | - | 4 | 851 | ||
AK300574.1 | - | 3 | 588 | ||
BC069270.1 | - | 3 | 690 | 5477442 |