Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025216.2 | NP_079492.2 | 2 | 635 | |
XM_011511929.2 | XP_011510231.1 | 3 | 243 | ||
XM_011511930.1 | XP_011510232.1 | 2 | 288 | ||
GenBank mRNA | AB059569.1 | - | 2 | 177 | |
AK024363.1 | - | 2 | 635 | ||
AK315081.1 | - | 2 | 288 | ||
AY009400.1 | - | 2 | 368 | ||
BC034352.1 | - | 2 | 294 | ||
BC040026.1 | - | 2 | 605 | ||
BC052234.1 | - | 2 | 646 | 5442628 |