Gene Transcripts

Gene Symbol
OR6N2
Entrez Gene ID
81442
Gene Name
olfactory receptor family 6 subfamily N member 2
Gene Aliases
OR1-23
Chromosome Location
Chr. 1: 158776682 - 158777635 on Build GRCh38
UniGene ID
Hs.554487
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005278.1 NP_001005278.1 1 814

View Full Product Details