Gene Transcripts

Gene Symbol
VWA9
Entrez Gene ID
81556
Gene Name
von Willebrand factor A domain containing 9
Gene Aliases
C15orf44
Chromosome Location
Chr. 15: 65578757 - 65611289 on Build GRCh38
UniGene ID
Hs.6686
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136043.2 NP_001129515.1 7 937
NM_001207058.1 NP_001193987.1 7 916
NM_001207059.1 NP_001193988.1 6 796
NR_045104.1 7 1080
NR_045105.2 7 1334
NR_045106.2 7 1241
NR_045901.1 6 972
XM_006720708.3 XP_006720771.1 7 1016
XM_006720709.3 XP_006720772.1 7 1081
XM_006720711.2 XP_006720774.1 6 940
XM_011522101.2 XP_011520403.1 7 762
XM_017022644.1 XP_016878133.1 7 1161
GenBank mRNA AK027475.1 - 7 922
AK293207.1 - 6 749
AK296134.1 - 7 1307
AK301033.1 - 7 779
AK301742.1 - 7 758
AL136662.1 - 7 1162
BC007991.2 - 7 907 3140504

View Full Product Details