Gene Transcripts

Gene Symbol
ANKRD13C
Entrez Gene ID
81573
Gene Name
ankyrin repeat domain 13C
Gene Aliases
dJ677H15.3
Chromosome Location
Chr. 1: 70259002 - 70354977 on Build GRCh38
UniGene ID
Hs.744989
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030816.4 NP_110443.3 13 1904
XM_005271234.3 XP_005271291.1 12 2046
XM_005271235.3 XP_005271292.1 11 1694
XM_006710929.3 XP_006710992.1 13 1873
XM_017002413.1 XP_016857902.1 11 1127
XM_017002414.1 XP_016857903.1 12 1209
XM_017002415.1 XP_016857904.1 12 1188
XM_017002416.1 XP_016857905.1 8 873
GenBank mRNA AK074187.1 - 5 528
AL136717.1 - 12 1800
BC028840.1 - 13 1893 4558748
BU622247.1 - 1 95

View Full Product Details