Gene Transcripts

Gene Symbol
KAZALD1
Entrez Gene ID
81621
Gene Name
Kazal type serine peptidase inhibitor domain 1
Gene Aliases
BONO1, FKSG28, FKSG40, IGFBP-rP10
Chromosome Location
Chr. 10: 101061141 - 101068141 on Build GRCh38
UniGene ID
Hs.534859
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001319303.1 NP_001306232.1 2 1215
NM_030929.4 NP_112191.2 2 1215
XM_005270194.3 XP_005270251.1 2 1316
XM_017016714.1 XP_016872203.1 1 294
XM_017016715.1 XP_016872204.1 1 294
GenBank mRNA AF333487.1 - 1 380
AK172862.1 - 2 466
AK172864.1 - 2 1214
AK225949.1 - 2 467
AY014271.1 - 1 380
AY359087.1 - 2 471
BC007758.2 - 2 616 4122366

View Full Product Details