Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001145646.1 | NP_001139118.1 | 5 | 589 | |
NM_031301.3 | NP_112591.2 | 6 | 712 | ||
GenBank mRNA | AA808355.1 | - | 1 | 276 | |
AB189172.1 | - | 5 | 529 | ||
AB197122.1 | - | 7 | 716 | ||
AF508794.1 | - | 6 | 652 | ||
AK291204.1 | - | 6 | 654 | ||
AK307725.1 | - | 6 | 774 | ||
AK310843.1 | - | 3 | 215 | ||
AL136671.1 | - | 6 | 658 | ||
AY358698.1 | - | 6 | 646 | ||
BC020905.1 | - | 6 | 654 | 4720647 |