Gene Transcripts
- Gene Symbol
- SPATA9
- Entrez Gene ID
- 83890
- Gene Name
- spermatogenesis associated 9
- Gene Aliases
- NYD-SP16
- Chromosome Location
- Chr. 5: 95652181 - 95731848 on Build GRCh38
- UniGene ID
- Hs.50499
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_031952.3 |
NP_114158.2
|
3, 4 |
528 |
|
NR_125327.1 |
|
3, 4 |
528 |
|
NR_125328.1 |
|
3, 4 |
503 |
|
XM_011543664.1 |
XP_011541966.1
|
3, 4 |
484 |
|
XM_017009948.1 |
XP_016865437.1
|
3, 4 |
509 |
|
XM_017009949.1 |
XP_016865438.1
|
8, 9 |
1065 |
|
XM_017009950.1 |
XP_016865439.1
|
7, 8 |
1047 |
|
GenBank mRNA |
AK292338.1 |
- |
3, 4 |
509 |
|
AK310260.1 |
- |
3, 4 |
484 |
|
AY027526.1 |
- |
3, 4 |
477 |
|
BC047333.1 |
- |
3, 4 |
514 |
5166271 |
View Full Product Details