Gene Transcripts

Gene Symbol
REG4
Entrez Gene ID
83998
Gene Name
regenerating family member 4
Gene Aliases
GISP, REG-IV, RELP
Chromosome Location
Chr. 1: 119794018 - 119811580 on Build GRCh38
UniGene ID
Hs.660883
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159352.1 NP_001152824.1 7 881
NM_032044.3 NP_114433.1 6 707
GenBank mRNA AF254415.1 - 6 622
AF345934.1 - 6 575
AY007243.1 - 6 563
AY126670.1 - 7 881
BC017089.2 - 6 579 3850358

View Full Product Details