Gene Transcripts

Gene Symbol
C3orf20
Entrez Gene ID
84077
Gene Name
chromosome 3 open reading frame 20
Gene Aliases
Chromosome Location
Chr. 3: 14673532 - 14773036 on Build GRCh38
UniGene ID
Hs.661452
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001184957.1 NP_001171886.1 11 1606
NM_001184958.1 NP_001171887.1 11 1711
NM_032137.4 NP_115513.4 11 2108
XM_006713341.2 XP_006713404.1 11 2108
XM_006713342.2 XP_006713405.1 11 1961
XM_006713343.2 XP_006713406.1 11 2108
XM_011534153.2 XP_011532455.1 11 3738
XM_011534154.2 XP_011532456.1 11 3173
XM_011534157.2 XP_011532459.1 7 885
GenBank mRNA AK131481.1 - 12 2314
AL136781.1 - 11 1526
AL834386.1 - 11 2060
BC030599.2 - 11 1669 4831210
BC038406.1 - 11 2028 5167835

View Full Product Details