Gene Transcripts

Gene Symbol
MYO18B
Entrez Gene ID
84700
Gene Name
myosin XVIIIB
Gene Aliases
KFS4
Chromosome Location
Chr. 22: 25742133 - 26063847 on Build GRCh38
UniGene ID
Hs.417959
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318245.1 NP_001305174.1 43 6953
NM_032608.6 NP_115997.5 43 6950
XM_011530458.2 XP_011528760.1 44 7089
XM_011530459.2 XP_011528761.1 44 7060
XM_011530460.2 XP_011528762.1 43 6933
XM_011530461.2 XP_011528763.1 44 7089
XM_011530464.2 XP_011528766.1 44 7086
XM_011530465.2 XP_011528767.1 43 6846
XM_017029012.1 XP_016884501.1 44 7089
XM_017029013.1 XP_016884502.1 44 7137
XM_017029014.1 XP_016884503.1 44 7086
XM_017029015.1 XP_016884504.1 43 6957
XM_017029016.1 XP_016884505.1 32 4213
XM_017029017.1 XP_016884506.1 42 6814
GenBank mRNA AB042648.2 - 39 5262
AB075376.1 - 43 6950
AJ310931.2 - 43 6941
AK095943.1 - 4 510
AK123936.1 - 4 540
AY077700.1 - 43 6863
BC144596.1 - 41 6605
BC144597.1 - 42 6747 9053127
BC150626.1 - 42 6747 9056996
CR749556.1 - 9 996

View Full Product Details