Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001048164.2 | NP_001041629.1 | 11 | 1831 | |
NM_032803.5 | NP_116192.4 | 11 | 1855 | ||
XM_017029912.1 | XP_016885401.1 | 10 | 2346 | ||
GenBank mRNA | AF320612.1 | - | 11 | 1806 | |
AK027447.1 | - | 9 | 1177 | ||
AK074865.1 | - | 11 | 1826 | ||
AK075014.1 | - | 11 | 1831 | ||
BC033816.1 | - | 11 | 1810 | 5206252 |