Gene Transcripts

Gene Symbol
SSH2
Entrez Gene ID
85464
Gene Name
slingshot protein phosphatase 2
Gene Aliases
SSH-2, SSH-2L
Chromosome Location
Chr. 17: 29625938 - 29930239 on Build GRCh38
UniGene ID
Hs.654754
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282129.1 NP_001269058.1 16 3268
NM_033389.3 NP_203747.2 15 2999
XM_005258058.3 XP_005258115.1 17 3161
XM_005258059.2 XP_005258116.1 14 3135
XM_005258060.3 XP_005258117.1 3 1728
XM_006722149.3 XP_006722212.1 14 6766
XM_011525402.2 XP_011523704.1 16 2996
XM_011525404.2 XP_011523706.1 14 3759
XM_011525405.2 XP_011523707.1 14 3807
XM_011525406.2 XP_011523708.1 13 3086
XM_011525407.1 XP_011523709.1 13 2831
XM_017025251.1 XP_016880740.1 16 3204
XM_017025252.1 XP_016880741.1 14 3065
GenBank mRNA AB051512.1 - 3 1817
AB099290.1 - 15 2959
AK091432.1 - 2 1388
AK296413.1 - 16 3133

View Full Product Details