Gene Transcripts

Gene Symbol
RSPH1
Entrez Gene ID
89765
Gene Name
radial spoke head 1 homolog
Gene Aliases
CT79, RSP44, RSPH10A, TSA2, TSGA2
Chromosome Location
Chr. 21: 42472486 - 42496354 on Build GRCh38
UniGene ID
Hs.661069
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286506.1 NP_001273435.1 6 658
NM_080860.3 NP_543136.1 7 772
XM_005261208.2 XP_005261265.1 5 412
XM_011529786.1 XP_011528088.1 6 592
GenBank mRNA AB006536.1 - 7 772
AB041016.1 - 7 709
AK057315.1 - 6 2223
AK314746.1 - 7 609
BC101519.1 - 7 651 8069025
BC113367.1 - 7 651 8322419
BG822861.1 - 492

View Full Product Details