Gene Transcripts

Gene Symbol
PLCZ1
Entrez Gene ID
89869
Gene Name
phospholipase C zeta 1
Gene Aliases
NYD-SP27, PLC-zeta-1, PLCzeta
Chromosome Location
Chr. 12: 18645529 - 18738059 on Build GRCh38
UniGene ID
Hs.743832
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033123.3 NP_149114.2 12 1710
NR_073075.1 12 1713
XM_005253519.2 XP_005253576.1 11 1826
XM_011520883.1 XP_011519185.1 9 1116
XM_017020176.1 XP_016875665.1 11 1826
XM_017020177.1 XP_016875666.1 12 1703
XM_017020178.1 XP_016875667.1 10 1676
XM_017020179.1 XP_016875668.1 11 1821
XM_017020180.1 XP_016875669.1 11 1715
XM_017020181.1 XP_016875670.1 11 1780
XM_017020182.1 XP_016875671.1 7 1479
XM_017020183.1 XP_016875672.1 8 1243
XM_017020184.1 XP_016875673.1 8 1171
XM_017020185.1 XP_016875674.1 7 1056
XM_017020186.1 XP_016875675.1 9 1078
GenBank mRNA AF532185.1 - 12 1635
AK097555.1 - 8 1058
AK097725.1 - 8 1176
AK292279.1 - 12 1708
AK314516.1 - 12 1667
AY035866.1 - 12 1672
BC125067.1 - 8 963 40117397
HY197954.1 - 4 443

View Full Product Details