Gene Transcripts

Gene Symbol
FATE1
Entrez Gene ID
89885
Gene Name
fetal and adult testis expressed 1
Gene Aliases
CT43, FATE
Chromosome Location
Chr. X: 151716036 - 151723194 on Build GRCh38
UniGene ID
Hs.301960
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033085.2 NP_149076.1 4 480
GenBank mRNA AF249872.1 - 4 480
AY101182.1 - 4 508
BC022064.2 - 4 481 4826440

View Full Product Details