Gene Transcripts
- Gene Symbol
- SPSB3
- Entrez Gene ID
- 90864
- Gene Name
- splA/ryanodine receptor domain and SOCS box containing 3
- Gene Aliases
- C16orf31, SSB3
- Chromosome Location
- Chr. 16: 1776712 - 1782605 on Build GRCh38
- UniGene ID
- Hs.592080
- Species
- Homo sapiens
- Gene Symbol
- EME2
- Entrez Gene ID
- 197342
- Gene Name
- essential meiotic structure-specific endonuclease subunit 2
- Gene Aliases
- SLX2B, gs125
- Chromosome Location
- Chr. 16: 1773222 - 1776714 on Build GRCh38
- UniGene ID
- Hs.7247
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK074080.2 |
- |
7 |
3293 |
|
View Full Product Details