Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_005268208.1 | XP_005268265.1 | 19 | 2472 | |
XM_005268209.3 | XP_005268266.1 | 19 | 2276 | ||
XM_017021765.1 | XP_016877254.1 | 19 | 2239 | ||
GenBank mRNA | AB082526.1 | - | 19 | 2408 | |
AK128693.1 | - | 18 | 2816 | ||
AL833036.1 | - | 1 | 276 | ||
AY048580.1 | - | 19 | 2302 | ||
AY080896.1 | - | 19 | 2314 | ||
BC001714.1 | - | 4 | 333 | ||
BC009336.2 | - | 4 | 344 | ||
BC093881.1 | - | 19 | 2383 | 7939726 | |
BC112101.1 | - | 19 | 2383 | 8327569 |