Gene Transcripts

Gene Symbol
SPECC1
Entrez Gene ID
92521
Gene Name
sperm antigen with calponin homology and coiled-coil domains 1
Gene Aliases
CYTSB, HCMOGT-1, HCMOGT1, NSP
Chromosome Location
Chr. 17: 20009301 - 20319026 on Build GRCh38
UniGene ID
Hs.431045
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033553.2 NP_001028725.1 4 1489
NM_001033554.2 NP_001028726.1 2 1398
NM_001033555.2 NP_001028727.1 2 1398
NM_001243438.1 NP_001230367.1 2 1398
NM_001243439.1 NP_001230368.1 4 1548
NM_152904.4 NP_690868.3 4 1489
XM_011524075.1 XP_011522377.1 3 1441
XM_017025317.1 XP_016880806.1 3 1441
XM_017025318.1 XP_016880807.1 3 1441
XM_017025319.1 XP_016880808.1 4 1583
XM_017025320.1 XP_016880809.1 4 1583
XM_017025321.1 XP_016880810.1 2 1398
XM_017025322.1 XP_016880811.1 3 1441
XM_017025323.1 XP_016880812.1 3 1441
XM_017025324.1 XP_016880813.1 3 1441
XM_017025325.1 XP_016880814.1 4 1583
GenBank mRNA AB041533.1 - 4 1489
AK295093.1 - 4 1521
AK295811.1 - 1 1025
AK298601.1 - 1 230
AK304186.1 - 2 1320
AK314341.1 - 4 1564
AY816326.1 - 2 1303
AY816327.1 - 2 1303
AY816328.1 - 4 1489
AY816329.1 - 4 1489
BC021123.1 - 2 1296 3614358
BC033618.1 - 1 513
BC050058.1 - 2 1233 5277450

View Full Product Details