Gene Transcripts
- Gene Symbol
- ORMDL3
- Entrez Gene ID
- 94103
- Gene Name
- ORMDL sphingolipid biosynthesis regulator 3
- Gene Aliases
- Chromosome Location
- Chr. 17: 39921041 - 39927804 on Build GRCh38
- UniGene ID
- Hs.514151
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001320801.1 |
NP_001307730.1
|
4, 5 |
1669 |
|
NM_001320802.1 |
NP_001307731.1
|
2, 3 |
507 |
|
NM_001320803.1 |
NP_001307732.1
|
2, 3 |
221 |
|
NM_139280.3 |
NP_644809.1
|
2, 3 |
512 |
|
XM_017025345.1 |
XP_016880834.1
|
2, 3 |
365 |
|
XM_017025346.1 |
XP_016880835.1
|
2, 3 |
278 |
|
XM_017025347.1 |
XP_016880836.1
|
2, 3 |
458 |
|
XM_017025348.1 |
XP_016880837.1
|
2, 3 |
204 |
|
GenBank mRNA |
AF373101.1 |
- |
2, 3 |
248 |
|
AF373102.1 |
- |
2, 3 |
318 |
|
AF395708.1 |
- |
2, 3 |
303 |
|
AK074811.1 |
- |
2, 3 |
309 |
|
AK075212.1 |
- |
2, 3 |
264 |
|
AK093063.1 |
- |
4, 5 |
1664 |
|
AX775843.1 |
- |
2, 3 |
266 |
|
BC017087.1 |
- |
2, 3 |
278 |
3850211 |
BC071833.1 |
- |
2, 3 |
257 |
6340437 |
DB042662.1 |
- |
2, 3 |
222 |
|
DB127807.1 |
- |
2, 3 |
340 |
|
HY004453.1 |
- |
2, 3 |
359 |
|
View Full Product Details