Gene Transcripts
- Gene Symbol
- IFT43
- Entrez Gene ID
- 112752
- Gene Name
- intraflagellar transport 43
- Gene Aliases
- C14orf179, CED3
- Chromosome Location
- Chr. 14: 75985753 - 76084073 on Build GRCh38
- UniGene ID
- Hs.532626
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001102564.1 |
NP_001096034.1
|
6, 7 |
412 |
|
NM_052873.2 |
NP_443105.2
|
5, 6 |
427 |
|
NR_045664.1 |
|
4, 5 |
332 |
|
NR_045665.1 |
|
6, 7 |
412 |
|
GenBank mRNA |
AK056735.1 |
- |
7 |
723 |
|
AK302944.1 |
- |
6, 7 |
395 |
|
BC010436.1 |
- |
5, 6 |
398 |
3608220 |
BI821833.1 |
- |
4, 5 |
308 |
|
CB853974.1 |
- |
6, 7 |
373 |
|
DB443802.1 |
- |
6, 7 |
411 |
|
DB497730.1 |
- |
6, 7 |
395 |
|
View Full Product Details