Gene Transcripts

Gene Symbol
MBD6
Entrez Gene ID
114785
Gene Name
methyl-CpG binding domain protein 6
Gene Aliases
Chromosome Location
Chr. 12: 57520710 - 57530148 on Build GRCh38
UniGene ID
Hs.524523
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_052897.3 NP_443129.3 6 1500
XM_005268617.1 XP_005268674.1 6 1553
XM_005268618.1 XP_005268675.1 6 1649
XM_005268620.2 XP_005268677.1 6 1553
XM_006719217.1 XP_006719280.1 6 1398
XM_006719218.1 XP_006719281.1 6 1386
XM_011537851.2 XP_011536153.1 6 2386
GenBank mRNA AB067474.1 - 3 1169
AK056399.1 - 6 1534
AK093078.1 - 1 395
AK296990.1 - 5 858
BC065530.1 - 6 1500 6152748

View Full Product Details