Gene Transcripts

Gene Symbol
CMTM5
Entrez Gene ID
116173
Gene Name
CKLF like MARVEL transmembrane domain containing 5
Gene Aliases
CKLFSF5
Chromosome Location
Chr. 14: 23376439 - 23379772 on Build GRCh38
UniGene ID
Hs.99272
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037288.1 NP_001032365.1 2 687
NM_001288746.1 NP_001275675.1 2 687
NM_138460.2 NP_612469.1 2 687
XM_017020953.1 XP_016876442.1 3 673
XM_017020954.1 XP_016876443.1 3 550
GenBank mRNA AF479262.2 - 2 688
AF527413.1 - 2 470
AK094840.1 - 1 1419
AY820135.1 - 2 243
BC013109.1 - 2 432 4214683

View Full Product Details