Gene Transcripts
- Gene Symbol
- WDR17
- Entrez Gene ID
- 116966
- Gene Name
- WD repeat domain 17
- Gene Aliases
- Chromosome Location
- Chr. 4: 176065834 - 176182828 on Build GRCh38
- UniGene ID
- Hs.532056
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_170710.4 |
NP_733828.2
|
13, 14 |
2078 |
|
NM_181265.3 |
NP_851782.3
|
12, 13 |
1955 |
|
XM_005262725.3 |
XP_005262782.1
|
11, 12 |
1601 |
|
XM_011531582.2 |
XP_011529884.1
|
12, 13 |
1943 |
|
XM_017007686.1 |
XP_016863175.1
|
12, 13 |
1931 |
|
XM_017007687.1 |
XP_016863176.1
|
13, 14 |
2016 |
|
XM_017007688.1 |
XP_016863177.1
|
13, 14 |
1933 |
|
XM_017007689.1 |
XP_016863178.1
|
12, 13 |
1930 |
|
XM_017007690.1 |
XP_016863179.1
|
13, 14 |
2016 |
|
XM_017007697.1 |
XP_016863186.1
|
12, 13 |
1932 |
|
GenBank mRNA |
AF492460.1 |
- |
13, 14 |
1931 |
|
View Full Product Details