Gene Transcripts
- Gene Symbol
- SIRPD
- Entrez Gene ID
- 128646
- Gene Name
- signal regulatory protein delta
- Gene Aliases
- PTPNS1L2, dJ576H24.4
- Chromosome Location
- Chr. 20: 1534251 - 1557697 on Build GRCh38
- UniGene ID
- Hs.729560
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK093083.1 |
- |
2 |
1347 |
|
BC033502.1 |
- |
2 |
201 |
5172531 |
View Full Product Details