Gene Transcripts

Gene Symbol
NUP35
Entrez Gene ID
129401
Gene Name
nucleoporin 35
Gene Aliases
MP-44, MP44, NP44, NUP53
Chromosome Location
Chr. 2: 183117490 - 183161684 on Build GRCh38
UniGene ID
Hs.180591
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001287584.1 NP_001274513.1 9 963
NM_001287585.1 NP_001274514.1 9 983
NM_138285.4 NP_612142.2 8 860
NR_109856.1 7 718
XM_006712254.2 XP_006712317.1 8 825
XM_011510576.2 XP_011508878.1 10 1091
XM_011510577.2 XP_011508879.1 10 1278
XM_017003308.1 XP_016858797.1 8 1016
GenBank mRNA AF411516.1 - 8 860
AF514993.1 - 8 817
AK289601.1 - 7 661
AK290760.1 - 8 772
AK298199.1 - 9 893
AK302350.1 - 9 932
BC035597.1 - 8 784
BC047029.1 - 8 847 5541023
BC061698.1 - 8 814 4132440

View Full Product Details