Gene Transcripts

Gene Symbol
AHSA2
Entrez Gene ID
130872
Gene Name
AHA1, activator of heat shock 90kDa protein ATPase homolog 2 (yeast)
Gene Aliases
Hch1
Chromosome Location
Chr. 2: 61177418 - 61191210 on Build GRCh38
UniGene ID
Hs.655602
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321300.1 NP_001308229.1 6, 7 1505
NM_001321301.1 NP_001308230.1 6, 7 2217
NM_001321302.1 NP_001308231.1 7, 8 925
NM_001321303.1 NP_001308232.1 6, 7 2203
NM_001321304.1 NP_001308233.1 4, 5 2928
NM_152392.4 NP_689605.1 5, 6 2101
XM_011532541.2 XP_011530843.1 7, 8 1161
XM_011532543.2 XP_011530845.1 8, 9 1445
XM_017003378.1 XP_016858867.1 8, 9 1120
GenBank mRNA AF542548.1 - 4, 5 302
AK092505.1 - 4, 5 1315
AK092817.1 - 5, 6 1505
AL833813.1 - 5, 6 1836
BC063789.1 - 5, 6 1990
BC113671.1 - 5, 6 1098 8322723
BC117278.1 - 5, 6 1098 40125829
BX505714.1 - 1, 2 62

View Full Product Details