Gene Transcripts

Gene Symbol
TAAR1
Entrez Gene ID
134864
Gene Name
trace amine associated receptor 1
Gene Aliases
TA1, TAR1, TRAR1
Chromosome Location
Chr. 6: 132644898 - 132646026 on Build GRCh38
UniGene ID
Hs.375030
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138327.2 NP_612200.1 1 107
GenBank mRNA AY180374.1 - 1 84
BC101825.1 - 1 107 8069331
BC112194.1 - 1 119 8327662

View Full Product Details