Gene Transcripts
- Gene Symbol
- FUNDC1
- Entrez Gene ID
- 139341
- Gene Name
- FUN14 domain containing 1
- Gene Aliases
- Chromosome Location
- Chr. X: 44523639 - 44542975 on Build GRCh38
- UniGene ID
- Hs.7549
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC042813.1 |
- |
2 |
291 |
|
View Full Product Details