Gene Transcripts

Gene Symbol
LYRM5
Entrez Gene ID
144363
Gene Name
LYR motif containing 5
Gene Aliases
Chromosome Location
Chr. 12: 25195216 - 25205051 on Build GRCh38
UniGene ID
Hs.209151
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001660.2 NP_001001660.2 2, 3 220
XM_005253319.4 XP_005253376.1 2, 3 223
XM_005253320.4 XP_005253377.1 3, 4 327
XM_017018850.1 XP_016874339.1 2, 3 1641
GenBank mRNA AK057730.1 - 2, 3 224
BC071769.1 - 2, 3 154 4246777
BC100276.1 - 2, 3 171 6178563
BG771497.1 - 2, 3 225

View Full Product Details