Gene Transcripts

Gene Symbol
FAM216B
Entrez Gene ID
144809
Gene Name
family with sequence similarity 216 member B
Gene Aliases
C13orf30
Chromosome Location
Chr. 13: 42781550 - 42791549 on Build GRCh38
UniGene ID
Hs.16970
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318932.1 NP_001305861.1 4 380
NM_182508.2 NP_872314.1 4 374
GenBank mRNA AK098238.1 - 4 315
BC093659.1 - 4 301 7939504
BC112237.1 - 4 301 8327705
DB221838.1 - 4 375

View Full Product Details