Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001317031.1 | NP_001303960.1 | 3 | 357 | |
NM_001317032.1 | NP_001303961.1 | 3 | 357 | ||
NM_001317033.1 | NP_001303962.1 | 3 | 470 | ||
NM_182513.3 | NP_872319.1 | 3 | 357 | ||
XM_011527702.1 | XP_011526004.1 | 3 | 327 | ||
GenBank mRNA | AK075287.1 | - | 3 | 327 | |
AK303266.1 | - | 3 | 465 | ||
AY456387.1 | - | 3 | 321 | ||
BC105037.1 | - | 3 | 353 | 8144040 | |
BC105039.1 | - | 3 | 353 | 8144042 | |
BQ061822.1 | - | 3 | 314 | ||
BQ216591.1 | - | 3 | 333 | ||
HY082333.1 | - | 3 | 360 |