Gene Transcripts

Gene Symbol
SIX5
Entrez Gene ID
147912
Gene Name
SIX homeobox 5
Gene Aliases
BOR2, DMAHP
Chromosome Location
Chr. 19: 45764785 - 45769239 on Build GRCh38
UniGene ID
Hs.43314
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175875.4 NP_787071.2 1, 2 1200
GenBank mRNA AK074826.1 - 1, 2 253
BU859227.1 - 614

View Full Product Details