Gene Transcripts

Gene Symbol
BROX
Entrez Gene ID
148362
Gene Name
BRO1 domain and CAAX motif containing
Gene Aliases
C1orf58
Chromosome Location
Chr. 1: 222712553 - 222735196 on Build GRCh38
UniGene ID
Hs.534965
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001288579.1 NP_001275508.1 4 652
NM_001288580.1 NP_001275509.1 4 358
NM_001288581.1 NP_001275510.1 4 358
NM_144695.3 NP_653296.2 4 652
XM_005273065.2 XP_005273122.2 4 288
XM_005273069.4 XP_005273126.1 3 524
XM_006711173.3 XP_006711236.1 5 721
XM_011509212.2 XP_011507514.1 4 711
XM_011509213.2 XP_011507515.1 3 596
XM_011509214.2 XP_011507516.1 5 521
XM_017000374.1 XP_016855863.1 6 846
XM_017000375.1 XP_016855864.1 3 597
XM_017000376.1 XP_016855865.1 4 649
GenBank mRNA AB276123.1 - 3 246
AK056983.1 - 4 641
AK293974.1 - 4 284
AK304445.1 - 4 303
AK315930.1 - 4 288
BC113635.1 - 4 389 8322687
BC113637.1 - 4 389 8322689
BC143811.1 - 4 389 9052326
DC418391.1 - 4 358

View Full Product Details