Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_015243.2 | NP_056058.2 | 2 | 124 | |
NM_017890.4 | NP_060360.3 | 2 | 124 | ||
NM_152564.4 | NP_689777.3 | 2 | 124 | ||
NM_181661.2 | NP_858047.2 | 2 | 124 | ||
NR_047582.1 | 2 | 124 | |||
XM_005250800.3 | XP_005250857.1 | 2 | 430 | ||
XM_005250801.4 | XP_005250858.1 | 2 | 6300 | ||
XM_006716510.3 | XP_006716573.1 | 2 | 116 | ||
XM_011516848.2 | XP_011515150.1 | 2 | 116 | ||
XM_011516849.2 | XP_011515151.1 | 2 | 116 | ||
XM_011516853.2 | XP_011515155.1 | 2 | 116 | ||
XM_011516859.2 | XP_011515161.1 | 2 | 116 | ||
XM_011516861.1 | XP_011515163.1 | 2 | 116 | ||
XM_011516865.1 | XP_011515167.1 | 2 | 116 | ||
XM_011516866.2 | XP_011515168.1 | 2 | 116 | ||
XM_017013109.1 | XP_016868598.1 | 2 | 116 | ||
GenBank mRNA | AJ608772.1 | - | 2 | 116 | |
AJ608773.1 | - | 2 | 116 | ||
AK000590.1 | - | 2 | 150 | ||
AK223122.1 | - | 2 | 150 | ||
AY223814.1 | - | 2 | 124 | ||
AY223815.1 | - | 2 | 124 | ||
AY223816.1 | - | 2 | 124 | ||
AY223817.1 | - | 2 | 124 | ||
AY223818.1 | - | 2 | 124 | ||
BC094749.1 | - | 2 | 80 | ||
BC132976.1 | - | 1 | 47 | ||
BC132978.1 | - | 1 | 47 |